(1-Methyl-1H-pyrrolo[2,3-b]pyridin-5-yl)boronic acid 97%

Supplier: Ambeed
Warning

A742648-1G A742648-250MG A742648-5G
76652-158EA 201.58 USD
76652-158 76652-160 76652-162
(1-Methyl-1H-pyrrolo[2,3-b]pyridin-5-yl)boronic acid 97%
(1-Methyl-1H-pyrrolo[2,3-b]pyridin-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MW: 175.98 g/mol
MDL Number: MFCD22394534
CAS Number: 1260536-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR