2-Chloro-4,5-dimethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A740428-100MG A740428-1G A740428-250MG
76652-098EA 44.56 USD
76652-098 76652-100 76652-102
2-Chloro-4,5-dimethoxyphenylboronic acid 98%
2-Chloro-4,5-dimethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BClO₄
MW: 216.43 g/mol
MDL Number: MFCD23379550
CAS Number: 950662-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR