Ethylenediaminetetraacetic acid disodium cobalt salt hydrate 98%

Supplier: Ambeed
Danger

Synonyms: Cobalt Disodium Ethylenediaminetetraacetate

A740167-25G A740167-5G
76652-094EA 14 USD
76652-094 76652-096
Ethylenediaminetetraacetic acid disodium cobalt salt hydrate 98%
Ethylenediaminetetraacetic acid disodium cobalt salt hydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₈
MW: 393.12 g/mol
MDL Number: MFCD00054448
CAS Number: 15137-09-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR