7-Bromo-1,8-naphthyridin-2(1H)-one 95%

Supplier: Ambeed
Danger

A740012-1G A740012-250MG
76652-092EA 456.19 USD
76652-092 77749-498
7-Bromo-1,8-naphthyridin-2(1H)-one 95%
7-Bromo-1,8-naphthyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MW: 225.0421 g/mol
MDL Number: MFCD31699217
CAS Number: 934559-24-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR