tert-Butyl 2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-4H,6H,7H-thieno[3,2-c]pyridine-5-carboxylate 98%

Supplier: Ambeed
Warning

A739997-100MG A739997-1G A739997-250MG A739997-5G
76652-084EA 36.08 USD
76652-084 76652-086 76652-088 76652-090
tert-Butyl 2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-4H,6H,7H-thieno[3,2-c]pyridine-5-carboxylate 98%
tert-Butyl 2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-4H,6H,7H-thieno[3,2-c]pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₈BNO₄S
MW: 365.2952 g/mol
MDL Number: MFCD27936521
CAS Number: 2246556-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR