2-Chloro-6-iodopyrazine 95%

Supplier: Ambeed
Warning

A739534-100MG A739534-250MG A739534-1G
76652-074EA 32.89 USD
76652-074 76652-076 77618-038
2-Chloro-6-iodopyrazine 95%
2-Chloro-6-iodopyrazine

Precaución: for Research Use Only

Formula: C₄H₂ClIN₂
MW: 240.4296 g/mol
MDL Number: MFCD26516149
CAS Number: 120531-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR