2,3-Dimethyl-7-nitroindole 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dimethyl-7-nitro-1H-indole

A739461-10G A739461-1G A739461-250MG A739461-5G
76652-068EA 192.03 USD
76652-068 76652-070 76652-072 77618-036
2,3-Dimethyl-7-nitroindole 98%
2,3-Dimethyl-7-nitroindole

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
MDL Number: MFCD00022710
CAS Number: 41018-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR