tert-Butyl ((3S,5R)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A736967-100MG A736967-1G
76652-054EA 82.76 USD
76652-054 77618-030
tert-Butyl ((3S,5R)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate 95%
tert-Butyl ((3S,5R)-5-(hydroxymethyl)pyrrolidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.28 g/mol
MDL Number: MFCD08704538
CAS Number: 1070295-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR