4-(2,4-Dimethylphenyl)-2-(1H-indol-3-yl)-4-oxobutanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Auxinole

A735100-100MG A735100-250MG A735100-50MG
76652-040EA 541.06 USD
76652-040 76652-042 76652-044
4-(2,4-Dimethylphenyl)-2-(1H-indol-3-yl)-4-oxobutanoic acid 98%
4-(2,4-Dimethylphenyl)-2-(1H-indol-3-yl)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₃
MW: 321.3698 g/mol
MDL Number: MFCD31657391
CAS Number: 86445-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR