5-NItro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95%
Supplier: Ambeed
|
Warning
|
A734196-100MG
A734196-1G
A734196-250MG
76652-028EA
24
USD
76652-028
76652-030
76652-032
5-NItro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95%
5-NItro-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Precaución: for Research Use Only
Formula:
C₁₃H₁₅BN₂O₄ |
MDL Number:
MFCD27935919 CAS Number: 1565857-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...