A-485 97%

Supplier: Ambeed
Warning

Synonyms: N-(4-Fluorobenzyl)-2-((R)-5-(3-methylureido)-2',4'-dioxo-2,3-dihydrospiro[indene-1,5'-oxazolidin]-3'-yl)-N-((S)-1,1,1-trifluoropropan-2-yl)acetamide

A732746-25MG A732746-10MG
76652-012EA 181.42 USD
76652-012 77617-980
A-485 97%
A-485

Precaución: for Research Use Only

Formula: C₂₅H₂₄F₄N₄O₅
MW: 536.48 g/mol
MDL Number: MFCD31619258
CAS Number: 1889279-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR