2-(Benzyloxy)-5-bromo-3-(trifluoromethyl)pyridine 96%

Supplier: Ambeed
Danger

A731275-100MG A731275-1G A731275-250MG
76652-002EA 13 USD
76652-002 76652-004 76652-006
2-(Benzyloxy)-5-bromo-3-(trifluoromethyl)pyridine 96%
2-(Benzyloxy)-5-bromo-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₃NO
MW: 332.1159 g/mol
MDL Number: MFCD17168431
CAS Number: 1824262-21-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR