1-(4-Bromophenyl)pyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A731116-100MG A731116-1G A731116-250MG
76651-996EA 42 USD
76651-996 76651-998 76652-000
1-(4-Bromophenyl)pyridin-2(1H)-one 95%
1-(4-Bromophenyl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₁H₈BrNO
MDL Number: MFCD30726957
CAS Number: 1845694-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR