3-Bromo-2-iodobenzonitrile 98%

Supplier: Ambeed
Warning

A730596-1G A730596-250MG
76651-992EA 121 USD
76651-992 76651-994
3-Bromo-2-iodobenzonitrile 98%
3-Bromo-2-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrIN
MDL Number: MFCD18391644
CAS Number: 450412-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR