1,2-distearoyl-sn-glycerol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Hydroxypropane-1,2-diyl distearate

A730537-250MG A730537-5G A730537-1G A730537-100MG
76651-990EA 72.15 USD
76651-990 77602-882 77598-858 77622-618
1,2-distearoyl-sn-glycerol 97%
1,2-distearoyl-sn-glycerol

Precaución: for Research Use Only

Formula: C₃₉H₇₆O₅
MW: 625.032 g/mol
MDL Number: MFCD00067473
CAS Number: 10567-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR