4,4,5,5-Tetramethyl-2-(4-((propylthio)methyl)phenyl)-1,3,2-dioxaborolane 98%
Supplier: Ambeed
|
Warning
|
A730162-100MG
A730162-1G
A730162-250MG
A730162-5G
76651-976EA
36.08
USD
76651-976
76651-978
76651-980
76651-982
4,4,5,5-Tetramethyl-2-(4-((propylthio)methyl)phenyl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(4-((propylthio)methyl)phenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₆H₂₅BO₂S MW: 292.2445 g/mol |
MDL Number:
MFCD18311858 CAS Number: 2246858-23-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...