(4-(Cyclohexylmethoxy)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A729949-100MG A729949-1G A729949-250MG A729949-5G
76651-954EA 37.14 USD
76651-954 76651-956 76651-958 76651-960
(4-(Cyclohexylmethoxy)phenyl)boronic acid 98%
(4-(Cyclohexylmethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₃
MW: 234.0992 g/mol
MDL Number: MFCD11183183
CAS Number: 938443-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR