Pyrazolo[1,5-a]pyrimidin-6-ylboronic acid 95%
Supplier: Ambeed
|
Warning
|
A729482-100MG
A729482-1G
A729482-250MG
76651-946EA
41
USD
76651-946
76651-948
76651-950
Pyrazolo[1,5-a]pyrimidin-6-ylboronic acid 95%
Pyrazolo[1,5-a]pyrimidin-6-ylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₆BN₃O₂ |
MDL Number:
MFCD27978523 CAS Number: 2828447-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...