3-(Tetrahydro-2H-pyran-4-yl)methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A729015-100MG A729015-1G A729015-250MG
76651-930EA 36.08 USD
76651-930 76651-932 76651-934
3-(Tetrahydro-2H-pyran-4-yl)methoxyphenylboronic acid 98%
3-(Tetrahydro-2H-pyran-4-yl)methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₄
MW: 236.0720 g/mol
MDL Number: MFCD17063326
CAS Number: 2096337-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR