(6-Acetylpyridin-3-yl)boronic acid 95%

Supplier: Ambeed
Warning

A728998-100MG A728998-1G A728998-250MG A728998-5G
76651-922EA 36.08 USD
76651-922 76651-924 76651-926 76651-928
(6-Acetylpyridin-3-yl)boronic acid 95%
(6-Acetylpyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₃
MW: 164.9543 g/mol
MDL Number: MFCD07374894
CAS Number: 2682066-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR