(4-(1-(Ethoxycarbonyl)cyclopropyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A728996-1G A728996-250MG A728996-5G
76651-916EA 210.06 USD
76651-916 76651-918 76651-920
(4-(1-(Ethoxycarbonyl)cyclopropyl)phenyl)boronic acid 97%
(4-(1-(Ethoxycarbonyl)cyclopropyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BO₄
MW: 234.06 g/mol
MDL Number: MFCD30741984
CAS Number: 1678539-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR