6,7-Dibromo-3H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one 97%

Supplier: Ambeed
Warning

A727898-100MG A727898-250MG
76651-902EA 74.27 USD
76651-902 76651-904
6,7-Dibromo-3H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one 97%
6,7-Dibromo-3H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one

Precaución: for Research Use Only

Formula: C₆H₃Br₂N₃O
MW: 292.9155 g/mol
MDL Number: MFCD31567431
CAS Number: 2173992-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR