2,4-Dichloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 96%

Supplier: Ambeed
Danger

A727119-100MG A727119-1G A727119-250MG
76651-896EA 48.81 USD
76651-896 76651-898 76651-900
2,4-Dichloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 96%
2,4-Dichloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BCl₂NO₂
MW: 273.9514 g/mol
MDL Number: MFCD18712633
CAS Number: 1622217-00-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR