4-(((1R,2R,4aR,5R,8aS)-2-((3-Carboxypropanoyl)oxy)-1,4a-dimethyl-6-methylene-5-((E)-2-(2-oxo-2,5-dihydrofuran-3-yl)vinyl)decahydronaphthalen-1-yl)methoxy)-4-oxobutanoic acid 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dehydroandrographolide succinate
A727104-100MG
A727104-250MG
A727104-1G
76651-892EA
59
USD
76651-892
76651-894
77598-854
4-(((1R,2R,4aR,5R,8aS)-2-((3-Carboxypropanoyl)oxy)-1,4a-dimethyl-6-methylene-5-((E)-2-(2-oxo-2,5-dihydrofuran-3-yl)vinyl)decahydronaphthalen-1-yl)methoxy)-4-oxobutanoic acid 99%
4-(((1R,2R,4aR,5R,8aS)-2-((3-Carboxypropanoyl)oxy)-1,4a-dimethyl-6-methylene-5-((E)-2-(2-oxo-2,5-dihydrofuran-3-yl)vinyl)decahydronaphthalen-1-yl)methoxy)-4-oxobutanoic acid
Precaución: for Research Use Only
Formula:
C₂₈H₃₆O₁₀ MW: 532.5795 g/mol |
MDL Number:
MFCD10566633 CAS Number: 786593-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...