(6-Chloro-2-methoxypyridin-3-yl)methanamine 95%

Supplier: Ambeed
Warning

A726298-100MG A726298-250MG
76651-888EA 119.89 USD
76651-888 76651-890
(6-Chloro-2-methoxypyridin-3-yl)methanamine 95%
(6-Chloro-2-methoxypyridin-3-yl)methanamine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MW: 172.6122 g/mol
MDL Number: MFCD28360473
CAS Number: 1784544-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR