N-Boc-N,N'-dimethylethylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: N1-BOC 1,2-Dimethyl-ethylenediamine, HCl

A726045-100MG A726045-250MG A726045-5G
76651-876EA 53 USD
76651-876 76651-878 76651-880
N-Boc-N,N'-dimethylethylamine hydrochloride 97%
N-Boc-N,N'-dimethylethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₀N₂O₂·ClH
MW: 224.728 g/mol
MDL Number: MFCD13184908
CAS Number: 202207-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR