6-(3-(Piperidin-1-yl)propoxy)-1,2,3,4-tetrahydroquinoline 97%

Supplier: Ambeed
Warning

A725799-100MG A725799-250MG
76651-872EA 96.55 USD
76651-872 76651-874
6-(3-(Piperidin-1-yl)propoxy)-1,2,3,4-tetrahydroquinoline 97%
6-(3-(Piperidin-1-yl)propoxy)-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₁₇H₂₆N₂O
MW: 274.4000 g/mol
MDL Number: MFCD31715275
CAS Number: 769920-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR