3,4,5-Tribromo-2-thiophenecarboxylic acid 95%

Supplier: Ambeed
Warning

A725712-1G A725712-250MG
76651-868EA 101 USD
76651-868 76651-870
3,4,5-Tribromo-2-thiophenecarboxylic acid 95%
3,4,5-Tribromo-2-thiophenecarboxylic acid

Precaución: for Research Use Only

Formula: C₅HBr₃O₂S
MDL Number: MFCD26939893
CAS Number: 53317-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR