(2S,4R)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (S,R,S)-AHPC-Me HCl

A725161-100MG A725161-10MG A725161-1G A725161-1MG A725161-250MG A725161-25MG A725161-50MG A725161-5G
76651-826EA 31.83 USD
76651-826 76651-828 76651-830 76651-832 76651-834 76651-836 76651-838 76651-840
(2S,4R)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride 98%
(2S,4R)-1-((S)-2-amino-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₃₃ClN₄O₃S
MW: 481.05 g/mol
MDL Number: MFCD30729675
CAS Number: 1948273-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR