1-(2-((4-Bromophenyl)amino)-2-oxoethyl)piperidine-4-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: BCI-121

A725132-100MG A725132-10MG A725132-1MG A725132-25MG A725132-50MG A725132-5MG A725132-250MG
76651-814EA 153.84 USD
76651-814 76651-816 76651-818 76651-820 76651-822 76651-824 77622-610
1-(2-((4-Bromophenyl)amino)-2-oxoethyl)piperidine-4-carboxamide 98%
1-(2-((4-Bromophenyl)amino)-2-oxoethyl)piperidine-4-carboxamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrN₃O₂
MW: 340.22 g/mol
MDL Number: MFCD02149239
CAS Number: 432529-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR