(R)-(4-Fluorophenyl)(8-methyl-3-(3-methyl-1,2,4-thiadiazol-5-yl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)methanone 98%

Supplier: Ambeed
Warning

A725085-10MG A725085-50MG A725085-5MG
76651-812EA 198 USD
76651-812 77783-786 77783-787
(R)-(4-Fluorophenyl)(8-methyl-3-(3-methyl-1,2,4-thiadiazol-5-yl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)methanone 98%
(R)-(4-Fluorophenyl)(8-methyl-3-(3-methyl-1,2,4-thiadiazol-5-yl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)methanone

Precaución: for Research Use Only

Formula: C₁₆H₁₅FN₆OS
MDL Number: MFCD30532641
CAS Number: 1629229-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR