BQR-695 98%

Supplier: Ambeed
Warning

Synonyms: NVP-BQR695, 2-((7-(3,4-dimethoxyphenyl)quinoxalin-2-yl)amino)-N-methylacetamide

A725053-10MG A725053-1MG A725053-50MG A725053-5MG A725053-100MG
76651-794EA 39 USD
76651-794 76651-796 76651-798 76651-800 77783-781
BQR-695 98%
BQR-695

Precaución: for Research Use Only

Formula: C₁₉H₂₀Cl₂N₄O₃
MDL Number: MFCD30533463
CAS Number: 1513879-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR