3-(Cyclopropylmethyl)-7-(4-phenylpiperidin-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
JNJ-42153605
A725030-10MG
A725030-1MG
A725030-5MG
76651-788EA
51.99
USD
76651-788
76651-790
76651-792
3-(Cyclopropylmethyl)-7-(4-phenylpiperidin-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine ≥98%
3-(Cyclopropylmethyl)-7-(4-phenylpiperidin-1-yl)-8-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridine
Precaución: for Research Use Only
Formula:
C₂₂H₂₃F₃N₄ MW: 400.44 g/mol |
MDL Number:
MFCD22199231 CAS Number: 1254977-87-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...