4'-((2-Butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl)methyl)-N-(4,5-dimethylisoxazol-3-yl)-2'-(ethoxymethyl)-[1,1'-biphenyl]-2-sulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: SParsentan

A725009-25MG A725009-100MG A725009-50MG
76651-786EA 199.45 USD
76651-786 77622-602 77622-604
4'-((2-Butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl)methyl)-N-(4,5-dimethylisoxazol-3-yl)-2'-(ethoxymethyl)-[1,1'-biphenyl]-2-sulfonamide 98%
4'-((2-Butyl-4-oxo-1,3-diazaspiro[4.4]non-1-en-3-yl)methyl)-N-(4,5-dimethylisoxazol-3-yl)-2'-(ethoxymethyl)-[1,1'-biphenyl]-2-sulfonamide

Precaución: for Research Use Only

Formula: C₃₂H₄₀N₄O₅S
MW: 592.7488 g/mol
MDL Number: MFCD16628036
CAS Number: 254740-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR