ASapiprant 98%

Supplier: Ambeed
Warning

Synonyms: 2-(5-(4-((4-Isopropoxyphenyl)sulphonyl)piperazin-1-yl)-2-(oxazol-2-yl)phenoxy)acetic acid

A724929-1MG A724929-5MG A724929-50MG
76651-782EA 20.16 USD
76651-782 76651-784 77622-600
ASapiprant 98%
ASapiprant

Precaución: for Research Use Only

Formula: C₂₄H₂₇N₃O₇S
MW: 501.55 g/mol
MDL Number: MFCD28902252
CAS Number: 932372-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR