1-Cyclopropyl-8-methyl-7-(5-methyl-6-(methylamino)pyridin-3-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Ozenoxacin

A724760-10MG A724760-1MG A724760-25MG A724760-5MG A724760-100MG A724760-250MG A724760-50MG A724760-1G
76651-766EA 112.46 USD
76651-766 76651-768 76651-770 76651-772 77598-834 77598-836 77598-838 77622-598
1-Cyclopropyl-8-methyl-7-(5-methyl-6-(methylamino)pyridin-3-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%
1-Cyclopropyl-8-methyl-7-(5-methyl-6-(methylamino)pyridin-3-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁N₃O₃
MW: 363.4097 g/mol
MDL Number: MFCD18633255
CAS Number: 245765-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR