1-Isopropyl-1'-(2-methyl-1H-benzo[d]imidazole-6-carbonyl)-4,6-dihydrospiro[indazole-5,4'-piperidin]-7(1H)-one 98+%

Supplier: Ambeed
Warning

Synonyms: PF-05175157

A724655-5MG A724655-10MG A724655-1MG A724655-50MG
76651-764EA 80.63 USD
76651-764 77622-592 77622-594 77622-596
1-Isopropyl-1'-(2-methyl-1H-benzo[d]imidazole-6-carbonyl)-4,6-dihydrospiro[indazole-5,4'-piperidin]-7(1H)-one 98+%
1-Isopropyl-1'-(2-methyl-1H-benzo[d]imidazole-6-carbonyl)-4,6-dihydrospiro[indazole-5,4'-piperidin]-7(1H)-one

Precaución: for Research Use Only

Formula: C₂₃H₂₇N₅O₂
MW: 405.49 g/mol
MDL Number: MFCD28502250
CAS Number: 1301214-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR