Methyl 2-(4-(2-(piperidin-1-yl)ethoxy)benzoyl)benzoate hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: Pitofenone HCl

A724539-10MG A724539-1MG A724539-25MG A724539-50MG A724539-5MG
76651-754EA 15 USD
76651-754 76651-756 76651-758 76651-760 76651-762
Methyl 2-(4-(2-(piperidin-1-yl)ethoxy)benzoyl)benzoate hydrochloride 98%
Methyl 2-(4-(2-(piperidin-1-yl)ethoxy)benzoyl)benzoate hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₆ClNO₄
MDL Number: MFCD01661944
CAS Number: 1248-42-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR