MK-3903 99+%

Supplier: Ambeed
Warning

Synonyms: MK3903, 5-((6-([1,1'-Biphenyl]-4-yl)-5-chloro-1H-benzo[d]imidazol-2-yl)oxy)-2-methylbenzoic acid, 5-((5-([1,1′-biphenyl]-4-yl)-6-chloro-1H-benzo[d]imidazol-2-yl)oxy)-2-methylbenzoic acid

A724442-10MG A724442-1MG A724442-25MG A724442-5MG
76651-738EA 103.97 USD
76651-738 76651-740 76651-742 76651-744
MK-3903 99+%
MK-3903

Precaución: for Research Use Only

Formula: C₂₇H₁₉ClN₂O₃
MW: 454.91 g/mol
MDL Number: MFCD22572341
CAS Number: 1219737-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR