5-(3-(Furan-2-carboxamido)benzamido)-2-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A724428-100MG A724428-10MG A724428-1G A724428-1MG A724428-250MG A724428-50MG A724428-5MG
76651-724EA 159.14 USD
76651-724 76651-726 76651-728 76651-730 76651-732 76651-734 76651-736
5-(3-(Furan-2-carboxamido)benzamido)-2-hydroxybenzoic acid 98%
5-(3-(Furan-2-carboxamido)benzamido)-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₄N₂O₆
MW: 366.3243 g/mol
MDL Number: MFCD07114226
CAS Number: 887686-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR