NIngetinib 98%

Supplier: Ambeed
Warning

Synonyms: N-(3-Fluoro-4-((7-(2-hydroxy-2-methylpropoxy)quinolin-4-yl)oxy)phenyl)-1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazole-4-carboxamide

A724397-10MG A724397-1MG A724397-25MG
76651-718EA 63.66 USD
76651-718 76651-720 76651-722
NIngetinib 98%
NIngetinib

Precaución: for Research Use Only

Formula: C₃₁H₂₉FN₄O₅
MW: 556.58 g/mol
MDL Number: MFCD28502048
CAS Number: 1394820-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR