1A-116 98%

Supplier: Ambeed
Warning

A724348-10MG A724348-1MG A724348-25MG A724348-5MG
76651-710EA 150 USD
76651-710 76651-712 76651-714 76651-716
1A-116 98%
1A-116

Precaución: for Research Use Only

Formula: C₁₆H₁₆F₃N₃
MDL Number: MFCD31619254
CAS Number: 1430208-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR