1-(4-Amino-2-((ethylamino)methyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol bis(2,2,2-trifluoroacetate) 98%

Supplier: Ambeed
Warning

A724317-10MG A724317-1MG A724317-25MG A724317-50MG A724317-5MG
76651-700EA 57 USD
76651-700 76651-702 76651-704 76651-706 76651-708
1-(4-Amino-2-((ethylamino)methyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol bis(2,2,2-trifluoroacetate) 98%
1-(4-Amino-2-((ethylamino)methyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol bis(2,2,2-trifluoroacetate)

Precaución: for Research Use Only

Formula: C₂₁H₂₅F₆N₅O₅
MDL Number: MFCD31619251
CAS Number: 1159840-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR