N-(3-Chloro-4-(((2-(methylamino)-6-(pyridin-2-yl)pyrimidin-4-yl)amino)methyl)phenyl)methanesulfonamide 98+%

Supplier: Ambeed
Warning

Synonyms: TC-G-1008

A724127-10MG A724127-1MG A724127-25MG A724127-50MG A724127-5MG
76651-666EA 143.23 USD
76651-666 76651-668 76651-670 76651-672 76651-674
N-(3-Chloro-4-(((2-(methylamino)-6-(pyridin-2-yl)pyrimidin-4-yl)amino)methyl)phenyl)methanesulfonamide 98+%
N-(3-Chloro-4-(((2-(methylamino)-6-(pyridin-2-yl)pyrimidin-4-yl)amino)methyl)phenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₁₈H₁₉ClN₆O₂S
MW: 418.9 g/mol
MDL Number: MFCD30182283
CAS Number: 1621175-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR