KKL-35 ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-N-(5-(4-fluorophenyl)-1,3,4-oxadiazol-2-yl)benzamide

A724020-10MG A724020-1MG A724020-5MG
76651-660EA 41.38 USD
76651-660 76651-662 76651-664
KKL-35 ≥98%
KKL-35

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClN₅O
MW: 265.70 g/mol
MDL Number: MFCD04438597
CAS Number: 865285-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR