3-(Benzo[d][1,3]dioxol-5-yl)-5-(3-bromophenyl)-1H-pyrazole 98%

Supplier: Ambeed
Warning

A723997-10MG A723997-1MG A723997-25MG A723997-50MG A723997-5MG A723997-250MG A723997-100MG
76651-650EA 62 USD
76651-650 76651-652 76651-654 76651-656 76651-658 77783-770 77783-769
3-(Benzo[d][1,3]dioxol-5-yl)-5-(3-bromophenyl)-1H-pyrazole 98%
3-(Benzo[d][1,3]dioxol-5-yl)-5-(3-bromophenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₆H₁₁BrN₂O₂
MDL Number: MFCD30146410
CAS Number: 882697-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR