2-(4-(4-Ethoxy-6-oxo-1,6-dihydropyridin-3-yl)-2-fluorophenyl)-N-(5-(1,1,1-trifluoro-2-methylpropan-2-yl)isoxazol-3-yl)acetamide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
GSK3179106
A723612-10MG
A723612-1MG
A723612-50MG
A723612-5MG
76651-618EA
84.88
USD
76651-618
76651-620
76651-622
76651-624
2-(4-(4-Ethoxy-6-oxo-1,6-dihydropyridin-3-yl)-2-fluorophenyl)-N-(5-(1,1,1-trifluoro-2-methylpropan-2-yl)isoxazol-3-yl)acetamide 98%
2-(4-(4-Ethoxy-6-oxo-1,6-dihydropyridin-3-yl)-2-fluorophenyl)-N-(5-(1,1,1-trifluoro-2-methylpropan-2-yl)isoxazol-3-yl)acetamide
Precaución: for Research Use Only
Formula:
C₂₂H₂₁F₄N₃O₄ MW: 467.41 g/mol |
MDL Number:
MFCD30747847 CAS Number: 1627856-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...