95%

Supplier: Ambeed
Warning

Synonyms: Cis-4-Phenylcyclohexan-1-ol

A723232-1G A723232-250MG A723232-5G
76651-606EA 145 USD
76651-606 76651-608 76651-610
95%
cis-4-Phenylcyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₆O
MW: 176.26
MDL Number: MFCD31692136
CAS Number: 7335-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR