5-(Difluoromethyl)-2-fluorobenzonitrile 95%

Supplier: Ambeed
Danger

A722620-100MG A722620-1G A722620-250MG A722620-50MG
76651-586EA 37 USD
76651-586 76651-588 76651-590 76651-592
5-(Difluoromethyl)-2-fluorobenzonitrile 95%
5-(Difluoromethyl)-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄F₃N
MDL Number: MFCD18395398
CAS Number: 934175-58-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR