(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)methanamine trifluoroacetic acid 95%

Supplier: Ambeed
Warning

A721965-100MG A721965-250MG A721965-1G
76651-582EA 116.7 USD
76651-582 76651-584 77622-562
(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)methanamine trifluoroacetic acid 95%
(4-(6-Methyl-1,2,4,5-tetrazin-3-yl)phenyl)methanamine trifluoroacetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₃N₅O₂
MW: 315.25 g/mol
MDL Number: MFCD31656825
CAS Number: 1466420-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR